Anti-VPS37C Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303042EA 550 EUR
ORIGTA303042
Anti-VPS37C Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: VPS37C (vacuolar protein sorting 37 homolog C (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR