Anti-COL7A1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303034EA 536 EUR
ORIGTA303034
Anti-COL7A1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-15µg/ml

Type: Primary
Antigen: COL7A1 (collagen, type VII, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR