Anti-NPY5R Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303031EA 547 EUR
ORIGTA303031
Anti-NPY5R Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: NPY5R (neuropeptide Y receptor Y5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR