Anti-CCKBR Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303029EA 547 EUR
ORIGTA303029
Anti-CCKBR Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000 Western Blot: 01-03µg/ml; Immunochemistry: 3µg/ml

Type: Primary
Antigen: CCKBR (cholecystokinin B receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR