Anti-MIF Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303028EA 547 EUR
ORIGTA303028
Anti-MIF Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1.5-3?g/ml; Western Blot: 0.01-0.03?g/ml

Type: Primary
Antigen: MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR