Anti-PCSK9 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303007EA 547 EUR
ORIGTA303007
Anti-PCSK9 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000 Western Blot: 02?g/ml; Immunochemistry: 2-6?g/ml

Type: Primary
Antigen: PCSK9 (proprotein convertase subtilisin/kexin type 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR