Anti-MYD88 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302999EA 539 EUR
ORIGTA302999
Anti-MYD88 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml; Immunochemistry: 2-5µg/ml

Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR