Anti-DYX1C1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302996EA 536 EUR
ORIGTA302996
Anti-DYX1C1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: DYX1C1 (dyslexia susceptibility 1 candidate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR