Anti-HSD17B10 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302984EA 547 EUR
ORIGTA302984
Anti-HSD17B10 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: HSD17B10 (hydroxysteroid (17-beta) dehydrogenase 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR