Anti-SYNGR2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302980EA 550 EUR
ORIGTA302980
Anti-SYNGR2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: SYNGR2 (synaptogyrin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR