Anti-TXNDC5 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302959EA 536 EUR
ORIGTA302959
Anti-TXNDC5 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-1µg/ml

Type: Primary
Antigen: TXNDC5 (thioredoxin domain containing 5 (endoplasmic reticulum))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR