Anti-FTCD Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302953EA 550 EUR
ORIGTA302953
Anti-FTCD Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-05µg/ml

Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR