Anti-CD2BP2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302949EA 547 EUR
ORIGTA302949
Anti-CD2BP2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: CD2BP2 (CD2 (cytoplasmic tail) binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR