Anti-CPEB1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302946EA 547 EUR
ORIGTA302946
Anti-CPEB1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: CPEB1 (cytoplasmic polyadenylation element binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR