Anti-ACTL6B Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302939EA 547 EUR
ORIGTA302939
Anti-ACTL6B Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ACTL6B (actin-like 6B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR