Anti-IFT74 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302937EA 547 EUR
ORIGTA302937
Anti-IFT74 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1µg/ml; Immunochemistry: 5-10µg/ml. Immunofluorescence: 2.5mg/ml

Type: Primary
Antigen: IFT74 (intraflagellar transport 74 homolog (Chlamydomonas))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR