Anti-USH1C Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302926EA 536 EUR
ORIGTA302926
Anti-USH1C Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml; Immunochemistry: 10µg/ml

Type: Primary
Antigen: USH1C (Usher syndrome 1C (autosomal recessive, severe))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR