Anti-MPP6 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302912EA 547 EUR
ORIGTA302912
Anti-MPP6 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: MPP6 (membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR