Anti-ATP6AP2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302894EA 547 EUR
ORIGTA302894
Anti-ATP6AP2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000. Western Blot: 0.25-1µg/ml

Type: Primary
Antigen: ATP6AP2 (ATPase, H+ transporting, lysosomal accessory protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR