Anti-ZBTB32 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302893EA 536 EUR
ORIGTA302893
Anti-ZBTB32 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 2-5µg/ml

Type: Primary
Antigen: ZBTB32 (zinc finger and BTB domain containing 32)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR