Anti-TCF19 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302889EA 536 EUR
ORIGTA302889
Anti-TCF19 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-2µg/ml

Type: Primary
Antigen: TCF19 (transcription factor 19)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR