Anti-FBXW2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302888EA 536 EUR
ORIGTA302888
Anti-FBXW2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-15µg/ml; Immunochemistry: 2-3µg/ml

Type: Primary
Antigen: FBXW2 (F-box and WD repeat domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR