Anti-FBXL2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302885EA 547 EUR
ORIGTA302885
Anti-FBXL2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: FBXL2 (F-box and leucine-rich repeat protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR