Anti-TRIM7 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302866EA 536 EUR
ORIGTA302866
Anti-TRIM7 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1µg/ml

Type: Primary
Antigen: TRIM7 (tripartite motif containing 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR