Anti-ARHGAP33 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302863EA 547 EUR
ORIGTA302863
Anti-ARHGAP33 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000 Western Blot: 25µg/ml

Type: Primary
Antigen: ARHGAP33 (Rho GTPase activating protein 33)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR