Anti-TRIM28 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302849EA 547 EUR
ORIGTA302849
Anti-TRIM28 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 02-1µg/ml

Type: Primary
Antigen: TRIM28 (tripartite motif containing 28)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR