Anti-HSPB8 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302835EA 536 EUR
ORIGTA302835
Anti-HSPB8 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000 Western Blot: 05-1µg/ml

Type: Primary
Antigen: HSPB8 (heat shock 22kDa protein 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR