Anti-ELF1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302810EA 547 EUR
ORIGTA302810
Anti-ELF1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: ELF1 (E74-like factor 1 (ets domain transcription factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR