Anti-KLF4 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302803EA 536 EUR
ORIGTA302803
Anti-KLF4 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR