Anti-ARPC1B Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302795EA 547 EUR
ORIGTA302795
Anti-ARPC1B Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 2-4µg/ml

Type: Primary
Antigen: ARPC1B (actin related protein 2/3 complex, subunit 1B, 41kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR