Anti-DBNL Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302793EA 536 EUR
ORIGTA302793
Anti-DBNL Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1/32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: DBNL (drebrin-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR