Anti-ADAM8 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302779EA 536 EUR
ORIGTA302779
Anti-ADAM8 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: ADAM8 (ADAM metallopeptidase domain 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR