Anti-DKK2 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA302770EA 550 EUR
ORIGTA302770
Anti-DKK2 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1: 8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: DKK2 (dickkopf WNT signaling pathway inhibitor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR