Anti-RAC2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302763EA 547 EUR
ORIGTA302763
Anti-RAC2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml; Immunochemistry: 03?g/ml

Type: Primary
Antigen: RAC2 (ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR