Anti-ELMO3 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA302757EA 547 EUR
ORIGTA302757
Anti-ELMO3 Goat polyclonal antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: ELMO3 (engulfment and cell motility 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR