Anti-DDAH1 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA302754EA 536 EUR
ORIGTA302754
Anti-DDAH1 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: DDAH1 (dimethylarginine dimethylaminohydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR