Anti-ATF7 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA302744EA 539 EUR
ORIGTA302744
Anti-ATF7 Goat polyclonal antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: ATF7 (activating transcription factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR