Anti-ORC4 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302715EA 547 EUR
ORIGTA302715
Anti-ORC4 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3 µg/ml

Type: Primary
Antigen: ORC4 (origin recognition complex, subunit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR