Anti-MAX Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302711EA 550 EUR
ORIGTA302711
Anti-MAX Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2 µg/ml

Type: Primary
Antigen: MAX (MYC associated factor X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR