Anti-SEC61A1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302702EA 547 EUR
ORIGTA302702
Anti-SEC61A1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4µg/ml

Type: Primary
Antigen: SEC61A1 (Sec61 alpha 1 subunit (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR