Anti-UBE2I Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302696EA 536 EUR
ORIGTA302696
Anti-UBE2I Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml; Immunochemistry: 1µg/ml

Type: Primary
Antigen: UBE2I (ubiquitin-conjugating enzyme E2I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR