Anti-SOCS1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302691EA 536 EUR
ORIGTA302691
Anti-SOCS1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2 µg/ml

Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR