Anti-RACGAP1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302684EA 547 EUR
ORIGTA302684
Anti-RACGAP1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 02-1µg/ml

Type: Primary
Antigen: RACGAP1 (Rac GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR