Anti-NCF1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302677EA 547 EUR
ORIGTA302677
Anti-NCF1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 02-2µg/ml

Type: Primary
Antigen: NCF1 (neutrophil cytosolic factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR