Anti-MMP7 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302676EA 536 EUR
ORIGTA302676
Anti-MMP7 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: MMP7 (matrix metallopeptidase 7 (matrilysin, uterine))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR