Anti-RPS6KB1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302669EA 536 EUR
ORIGTA302669
Anti-RPS6KB1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.05-0.1 ?g/ml; Immunochemistry: 2-4 ?g/?l

Type: Primary
Antigen: RPS6KB1 (ribosomal protein S6 kinase, 70kDa, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR