Anti-HSPA8 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302666EA 536 EUR
ORIGTA302666
Anti-HSPA8 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR