Anti-FXR1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302663EA 550 EUR
ORIGTA302663
Anti-FXR1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: FXR1 (fragile X mental retardation, autosomal homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR