Anti-FOXP3 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302662EA 547 EUR
ORIGTA302662
Anti-FOXP3 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Immunochemistry: 03-1 µg/ml

Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR