Anti-PTGS1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302656EA 536 EUR
ORIGTA302656
Anti-PTGS1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: PTGS1 (prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR